3-Ethyl-4-fluorophenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

47342-1G 47342-500MG 47342-5G
76916-510EA 468.92 USD
76916-510 76916-512 76916-514
3-Ethyl-4-fluorophenylboronic acid ≥95%
3-Ethyl-4-fluorophenylboronic acid
Formula: C₈H₁₀BFO₂
MW: 167.973 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22491271
CAS Number: 960235-01-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR