3-Chloro-2-fluoro-5-iodobenzoic acid ≥97% research grade

Supplier: AOBChem USA

Warning

47268-1G 47268-25G 47268-500MG 47268-5G
76916-496EA 129.43 USD
76916-496 76916-498 76916-500 76916-502
3-Chloro-2-fluoro-5-iodobenzoic acid ≥97% research grade
3-Chloro-2-fluoro-5-iodobenzoic acid
Formula: C₇H₃ClFIO₂
Storage Temperature: Refrigerator
MDL Number: MFCD24107275
CAS Number: 1000162-09-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR