4'-Bromobiphenyl-4-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

46966-1G 46966-25G 46966-5G
76916-482EA 66.84 USD
76916-482 76916-484 76916-486
4'-Bromobiphenyl-4-carboxylic acid ≥97%
4'-Bromobiphenyl-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₉BrO₂
Storage Temperature: Ambient
MDL Number: MFCD00452920
CAS Number: 5731-11-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR