3-(Benzyloxy)-5-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

46739-1G 46739-25G 46739-500MG 46739-5G
76916-466EA 345.86 USD
76916-466 76916-468 76916-470 76916-472
3-(Benzyloxy)-5-methoxybenzaldehyde ≥97%
3-(Benzyloxy)-5-methoxybenzaldehyde
Formula: C₁₅H₁₄O₃
Storage Temperature: Refrigerator
CAS Number: 50637-29-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR