4-Bromo-1-isopropyl-2-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

46409-1G 46409-25G 46409-500MG 46409-5G
76916-450EA 250.38 USD
76916-450 76916-452 76916-454 76916-456
4-Bromo-1-isopropyl-2-methoxybenzene ≥97%
4-Bromo-1-isopropyl-2-methoxybenzene
Formula: C₁₀H₁₃BrO
MW: 229.1136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28356548
CAS Number: 1369775-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR