3-Bromo-N-cyclopropyl-5-fluorobenzamide ≥97%

Supplier: AOBChem USA

Warning

45938-1G 45938-25G 45938-5G
76916-398EA 106.09 USD
76916-398 76916-400 76916-402
3-Bromo-N-cyclopropyl-5-fluorobenzamide ≥97%
3-Bromo-N-cyclopropyl-5-fluorobenzamide
Formula: C₁₀H₉BrFNO
MW: 258.087 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13306978
CAS Number: 1147712-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR