3-Bromo-2-ethoxy-5-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

45923-1G 45923-25G 45923-500MG 45923-5G
76916-382EA 290.69 USD
76916-382 76916-384 76916-386 76916-388
3-Bromo-2-ethoxy-5-methylbenzoic acid ≥97%
3-Bromo-2-ethoxy-5-methylbenzoic acid
Formula: C₁₀H₁₁BrO₃
MW: 259.097 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707143
CAS Number: 1535334-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR