3,5-Dichloro-2-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

45656-25G 45656-5G
76916-374EA 537.88 USD
76916-374 76916-376
3,5-Dichloro-2-methoxypyridine ≥97%
3,5-Dichloro-2-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅Cl₂NO
MW: 178.016 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185543
CAS Number: 13472-58-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR