3-Ethoxy-4-fluorobenzoic acid ≥97% research grade

Supplier: AOBChem USA

Warning

45534-1G 45534-25G 45534-500MG 45534-5G
76916-350EA 332.07 USD
76916-350 76916-352 76916-354 76916-356
3-Ethoxy-4-fluorobenzoic acid ≥97% research grade
3-Ethoxy-4-fluorobenzoic acid
Formula: C₉H₉FO₃
Storage Temperature: Ambient
MDL Number: MFCD18375410
CAS Number: 445026-29-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR