4-Bromo-2-chloro-5-(trifluoromethyl)benzoic acid ≥97% research grade

Supplier: AOBChem USA

Warning

45477-1G 45477-25G 45477-500MG 45477-5G
76916-334EA 260.99 USD
76916-334 76916-336 76916-338 76916-340
4-Bromo-2-chloro-5-(trifluoromethyl)benzoic acid ≥97% research grade
4-Bromo-2-chloro-5-(trifluoromethyl)benzoic acid
Formula: C₈H₃BrClF₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD32707119
CAS Number: 2092139-72-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR