3,5-Dibromo-4-(dimethoxymethyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

45427-25G 45427-5G
76916-322EA 310.85 USD
76916-322 76916-324
3,5-Dibromo-4-(dimethoxymethyl)pyridine ≥95%
3,5-Dibromo-4-(dimethoxymethyl)pyridine
Formula: C₈H₉Br₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD32707117
CAS Number: 2413441-27-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR