4-(4-Formylphenoxy)phenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

45174-1G 45174-25G 45174-500MG 45174-5G
76916-310EA 274.78 USD
76916-310 76916-312 76916-314 76916-316
4-(4-Formylphenoxy)phenylboronic acid ≥95%
4-(4-Formylphenoxy)phenylboronic acid
Formula: C₁₃H₁₁BO₄
Storage Temperature: Ambient
MDL Number: MFCD29047677
CAS Number: 1416252-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR