4'-Borono-[1,1'-biphenyl]-4-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

44918-1G 44918-25G 44918-5G
76916-298EA 114.58 USD
76916-298 76916-300 76916-302
4'-Borono-[1,1'-biphenyl]-4-carboxylic acid ≥97%
4'-Borono-[1,1'-biphenyl]-4-carboxylic acid
Formula: C₁₃H₁₁BO₄
MW: 242.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29077670
CAS Number: 872341-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR