3-Bromo-2-fluoro-6-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

44763-1G 44763-25G 44763-5G
76916-284EA 96.55 USD
76916-284 76916-286 76916-288
3-Bromo-2-fluoro-6-methylphenylboronic acid ≥97%
3-Bromo-2-fluoro-6-methylphenylboronic acid
Formula: C₇H₇BBrFO₂
MW: 232.84 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28515375
CAS Number: 2246650-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR