4-(Boc-amino)-2-fluorobenzeneboronic acid pinacol ester ≥95% research grade

Supplier: AOBChem USA

Warning

Synonyms: tert-Butyl (3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate, 4-(BOC-Amino)-2-fluorophenylboronic acid pinacol ester

44185-1G 44185-25G 44185-5G
76916-270EA 62.6 USD
76916-270 76916-272 76916-274
4-(Boc-amino)-2-fluorobenzeneboronic acid pinacol ester ≥95% research grade
4-(Boc-amino)-2-fluorobenzeneboronic acid pinacol ester
Formula: C₁₇H₂₅BFNO₄
Storage Temperature: Ambient
MDL Number: MFCD18383441
CAS Number: 1256256-45-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR