3,5-Dichloro-4-fluorophenylboronic acid ≥97% research grade
Supplier: AOBChem USA
|
Warning
|
42102-1G
42102-25G
42102-500MG
42102-5G
76916-254EA
147.47
USD
76916-254
76916-256
76916-258
76916-260
3,5-Dichloro-4-fluorophenylboronic acid ≥97% research grade
3,5-Dichloro-4-fluorophenylboronic acid
Formula:
C₆H₄BCl₂FO₂ MW: 208.81 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18837599 CAS Number: 1646614-31-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...