3-Methoxy-5-(methoxycarbonyl)phenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Methyl 3-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 3-Methoxy-5-(methoxycarbonyl)phenylboronic acid, pinacol ester

41601-1G 41601-25G 41601-500MG 41601-5G
76916-238EA 381.93 USD
76916-238 76916-240 76916-242 76916-244
3-Methoxy-5-(methoxycarbonyl)phenylboronic acid pinacol ester ≥97%
3-Methoxy-5-(methoxycarbonyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₁BO₅
MW: 292.135 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12026074
CAS Number: 889654-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR