3-Ethoxy-4-(trifluoromethoxy)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

40909-1G 40909-25G 40909-500MG 40909-5G
76916-206EA 381.93 USD
76916-206 76916-208 76916-210 76916-212
3-Ethoxy-4-(trifluoromethoxy)phenylboronic acid ≥97%
3-Ethoxy-4-(trifluoromethoxy)phenylboronic acid
Formula: C₉H₁₀BF₃O₄
MW: 249.98 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21332933
CAS Number: 1701449-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR