(4'-(Trifluoromethyl)-[1,1'-biphenyl]-3-yl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 3-[4-(trifluoromethyl)phenyl]phenylboronic acid

40634-1G 40634-500MG 40634-5G
76916-180EA 542.12 USD
76916-180 76916-182 76916-184
(4'-(Trifluoromethyl)-[1,1'-biphenyl]-3-yl)boronic acid ≥97% research grade
(4'-(Trifluoromethyl)-[1,1'-biphenyl]-3-yl)boronic acid
Formula: C₁₃H₁₀BF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD08701741
CAS Number: 1107603-45-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR