4-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy)benzonitrile ≥97%
Supplier: AOBChem USA
|
Warning
|
40450-1G
40450-25G
40450-5G
76916-168EA
76.39
USD
76916-168
76916-170
76916-172
4-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy)benzonitrile ≥97%
4-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy)benzonitrile
Formula:
C₁₉H₂₀BNO₃ MW: 321.178 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD20527107 CAS Number: 2096998-25-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...