3-Chloro-4-fluoro-5-methylphenylboronic acid, pinacol ester ≥97%
Supplier: AOBChem USA
|
Warning
|
40278-1G
40278-25G
40278-500MG
40278-5G
76916-140EA
381.93
USD
76916-140
76916-142
76916-144
76916-146
3-Chloro-4-fluoro-5-methylphenylboronic acid, pinacol ester ≥97%
3-Chloro-4-fluoro-5-methylphenylboronic acid, pinacol ester
Formula:
C₁₃H₁₇BClFO₂ Storage Temperature: Ambient |
MDL Number:
MFCD20231468 CAS Number: 1402238-26-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...