3-Carboxy-4-(trifluoromethoxy)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

40254-1G 40254-25G 40254-500MG 40254-5G
76916-124EA 236.59 USD
76916-124 76916-126 76916-128 76916-130
3-Carboxy-4-(trifluoromethoxy)phenylboronic acid ≥97%
3-Carboxy-4-(trifluoromethoxy)phenylboronic acid
Formula: C₈H₆BF₃O₅
Storage Temperature: Refrigerator
MDL Number: MFCD20231437
CAS Number: 2096329-67-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR