(3-Methyl-5-propoxyphenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 3-Methyl-5-propoxyphenylboronic acid

39751-1G 39751-25G 39751-500MG 39751-5G
76916-080EA 155.96 USD
76916-080 76916-082 76916-084 76916-086
(3-Methyl-5-propoxyphenyl)boronic acid ≥97% research grade
(3-Methyl-5-propoxyphenyl)boronic acid
Formula: C₁₀H₁₅BO₃
Storage Temperature: Ambient
MDL Number: MFCD16295067
CAS Number: 1256346-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR