3-Isopropoxy-5-(trifluoromethoxy)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (3-Isopropoxy-5-(trifluoromethoxy)phenyl)boronic acid

39747-1G 39747-25G 39747-5G
76916-068EA 102.91 USD
76916-068 76916-070 76916-072
3-Isopropoxy-5-(trifluoromethoxy)phenylboronic acid ≥97%
3-Isopropoxy-5-(trifluoromethoxy)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂BF₃O₄
MW: 264.0100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16295063
CAS Number: 1256346-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR