3-Isopropoxy-5-methylphenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: (3-Isopropoxy-5-methylphenyl)boronic acid

39712-1G 39712-25G 39712-500MG 39712-5G
76916-054EA 126.25 USD
76916-054 76916-056 76916-058 76916-060
3-Isopropoxy-5-methylphenylboronic acid ≥97% research grade
3-Isopropoxy-5-methylphenylboronic acid
Formula: C₁₀H₁₅BO₃
Storage Temperature: Ambient
MDL Number: MFCD16295054
CAS Number: 1256345-76-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR