3-Dimethylaminopyridine-5-boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

39595-1G 39595-25G 39595-500MG 39595-5G
76916-042EA 350.1 USD
76916-042 76916-044 76916-046 76916-048
3-Dimethylaminopyridine-5-boronic acid ≥97% research grade
3-Dimethylaminopyridine-5-boronic acid
Formula: C₇H₁₁BN₂O₂
MW: 165.985 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12964565
CAS Number: 1018680-09-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR