4-(Isobutylthio)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

39467-25G 39467-5G
76916-030EA 183.54 USD
76916-030 76916-032
4-(Isobutylthio)phenylboronic acid ≥97%
4-(Isobutylthio)phenylboronic acid
Formula: C₁₀H₁₅BO₂S
MW: 210.101 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12546543
CAS Number: 1217500-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR