3-Bromo-2,5-dimethylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

38703-1G 38703-25G 38703-500MG 38703-5G
76915-984EA 439.22 USD
76915-984 76915-986 76915-988 76915-990
3-Bromo-2,5-dimethylbenzoic acid ≥97%
3-Bromo-2,5-dimethylbenzoic acid
Formula: C₉H₉BrO₂
MW: 229.071 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD26403724
CAS Number: 1255209-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR