3-Methoxy-4-propoxybenzaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

38324-25G 38324-5G
76915-968EA 598.35 USD
76915-968 76915-970
3-Methoxy-4-propoxybenzaldehyde ≥97% research grade
3-Methoxy-4-propoxybenzaldehyde
Formula: C₁₁H₁₄O₃
Storage Temperature: Refrigerator
MDL Number: MFCD00189329
CAS Number: 57695-98-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR