3-Iodo-2-methylimidazo[1,2-a]pyridine ≥95%

Supplier: AOBChem USA

Warning

38292-1G 38292-25G 38292-5G
76915-958EA 106.09 USD
76915-958 76915-960 76915-962
3-Iodo-2-methylimidazo[1,2-a]pyridine ≥95%
3-Iodo-2-methylimidazo[1,2-a]pyridine
Formula: C₈H₇IN₂
MW: 258.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16250856
CAS Number: 1204527-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR