4-(Hydroxymethyl)-2-methoxybenzonitrile ≥97%
Supplier: AOBChem USA
|
Warning
|
38247-1G
38247-500MG
38247-5G
76915-944EA
558.04
USD
76915-944
76915-946
76915-948
4-(Hydroxymethyl)-2-methoxybenzonitrile ≥97%
4-(Hydroxymethyl)-2-methoxybenzonitrile
Formula:
C₉H₉NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD18394233 CAS Number: 210037-77-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...