4,6-Dibromo-2-fluoro-3-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

38158-1G 38158-25G 38158-500MG 38158-5G
76915-918EA 260.99 USD
76915-918 76915-920 76915-922 76915-924
4,6-Dibromo-2-fluoro-3-methoxybenzoic acid ≥97%
4,6-Dibromo-2-fluoro-3-methoxybenzoic acid
Formula: C₈H₅Br₂FO₃
MW: 327.93 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32707068
CAS Number: 2413441-19-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR