3-Bromo-N,N-dimethyl-5-nitrobenzamide ≥97%

Supplier: AOBChem USA

Warning

38069-1G 38069-25G 38069-5G
76915-908EA 88.06 USD
76915-908 76915-910 76915-912
3-Bromo-N,N-dimethyl-5-nitrobenzamide ≥97%
3-Bromo-N,N-dimethyl-5-nitrobenzamide

Precaución: for Research Use Only

Formula: C₉H₉BrN₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD09258761
CAS Number: 929000-26-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR