3-Bromo-N-methyl-5-nitrobenzamide ≥97%

Supplier: AOBChem USA

Warning

38063-1G 38063-25G 38063-5G
76915-898EA 88.06 USD
76915-898 76915-900 76915-902
3-Bromo-N-methyl-5-nitrobenzamide ≥97%
3-Bromo-N-methyl-5-nitrobenzamide

Precaución: for Research Use Only

Formula: C₈H₇BrN₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD09475916
CAS Number: 90050-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR