3-Bromo-9-ethylcarbazole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Bromo-9-ethyl-9H-carbazole

38046-25G 38046-5G
76915-886EA 471.04 USD
76915-886 76915-888
3-Bromo-9-ethylcarbazole ≥97%
3-Bromo-9-ethylcarbazole

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrN
MW: 274.1558 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00092392
CAS Number: 57102-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR