3,5-Dimethyl-4-propoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

38004-1G 38004-25G 38004-500MG 38004-5G
76915-874EA 263.11 USD
76915-874 76915-876 76915-878 76915-880
3,5-Dimethyl-4-propoxybenzoic acid ≥97%
3,5-Dimethyl-4-propoxybenzoic acid
Formula: C₁₂H₁₆O₃
MW: 208.254 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11188409
CAS Number: 1039885-39-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR