3-Bromo-5-chloro-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

37942-1G 37942-25G 37942-500MG 37942-5G
76915-842EA 350.1 USD
76915-842 76915-844 76915-846 76915-848
3-Bromo-5-chloro-N-methylbenzamide ≥97%
3-Bromo-5-chloro-N-methylbenzamide
Formula: C₈H₇BrClNO
MW: 248.504 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD26610232
CAS Number: 1532970-00-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR