3-(5-Chloro-6-methoxypyridin-3-yl)acrylic acid ≥97%

Supplier: AOBChem USA

Warning

37875-1G 37875-25G 37875-500MG 37875-5G
76915-828EA 421.18 USD
76915-828 76915-830 76915-832 76915-834
3-(5-Chloro-6-methoxypyridin-3-yl)acrylic acid ≥97%
3-(5-Chloro-6-methoxypyridin-3-yl)acrylic acid
Formula: C₉H₈ClNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD32632266
CAS Number: 2360396-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR