3,4-Dichloro-2-fluorophenol ≥97% research grade

Supplier: AOBChem USA

Warning

37604-1G 37604-25G 37604-500MG 37604-5G
76915-786EA 346.92 USD
76915-786 76915-788 76915-790 76915-792
3,4-Dichloro-2-fluorophenol ≥97% research grade
3,4-Dichloro-2-fluorophenol
Formula: C₆H₃Cl₂FO
Storage Temperature: Refrigerator
MDL Number: MFCD24719620
CAS Number: 934180-44-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR