4,6-Difluoro-3-iodo-1H-indazole ≥97%
Supplier: AOBChem USA
|
Warning
|
37554-1G
37554-25G
37554-500MG
37554-5G
76915-770EA
242.95
USD
76915-770
76915-772
76915-774
76915-776
4,6-Difluoro-3-iodo-1H-indazole ≥97%
4,6-Difluoro-3-iodo-1H-indazole
Formula:
C₇H₃F₂IN₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD07781823 CAS Number: 887567-79-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...