3-Fluoro-2-(methoxymethyl)-6-methylbenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

37193-1G 37193-500MG 37193-5G
76915-734EA 749 USD
76915-734 76915-736 76915-738
3-Fluoro-2-(methoxymethyl)-6-methylbenzaldehyde ≥95%
3-Fluoro-2-(methoxymethyl)-6-methylbenzaldehyde
Formula: C₁₀H₁₁FO₂
MW: 182.192 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632157
CAS Number: 2379322-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR