4-(3-Methylphenyl)phenol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3'-Methyl-[1,1'-biphenyl]-4-ol

37102-1G 37102-25G 37102-500MG 37102-5G
76915-720EA 124.13 USD
76915-720 76915-722 76915-724 76915-726
4-(3-Methylphenyl)phenol ≥97%
4-(3-Methylphenyl)phenol

Precaución: for Research Use Only

Formula: C₁₃H₁₂O
MW: 184.234 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01910124
CAS Number: 191724-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR