4-(3-Bromophenyl)-1,3-thiazol-2-amine ≥97%
Supplier: AOBChem USA
|
Warning
|
37054-25G
37054-5G
76915-710EA
559.1
USD
76915-710
76915-712
4-(3-Bromophenyl)-1,3-thiazol-2-amine ≥97%
4-(3-Bromophenyl)-1,3-thiazol-2-amine
Formula:
C₉H₇BrN₂S Storage Temperature: Ambient |
MDL Number:
MFCD00170238 CAS Number: 105512-81-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...