3,5-Dichloro-4-isopropoxyphenol ≥97%

Supplier: AOBChem USA

Warning

37028-1G 37028-25G 37028-500MG 37028-5G
76915-696EA 320.4 USD
76915-696 76915-698 76915-700 76915-702
3,5-Dichloro-4-isopropoxyphenol ≥97%
3,5-Dichloro-4-isopropoxyphenol
Formula: C₉H₁₀Cl₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD23111016
CAS Number: 1394927-31-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR