3,5-Difluoro-4-(trimethylsilyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

36820-1G 36820-500MG 36820-5G
76915-686EA 733.09 USD
76915-686 76915-688 76915-690
3,5-Difluoro-4-(trimethylsilyl)benzoic acid ≥97%
3,5-Difluoro-4-(trimethylsilyl)benzoic acid
Formula: C₁₀H₁₂F₂O₂Si
Storage Temperature: Ambient
MDL Number: MFCD30178499
CAS Number: 872545-56-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR