4-Bromo-2-(methoxymethyl)-1-(trifluoromethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

36734-1G 36734-25G 36734-500MG 36734-5G
76915-656EA 253.56 USD
76915-656 76915-658 76915-660 76915-662
4-Bromo-2-(methoxymethyl)-1-(trifluoromethoxy)benzene ≥97%
4-Bromo-2-(methoxymethyl)-1-(trifluoromethoxy)benzene
Formula: C₉H₈BrF₃O₂
MW: 285.058 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632077
CAS Number: 2379321-82-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR