3-Fluoro-2-methoxy-5-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

36674-1G 36674-500MG 36674-5G
76915-624EA 510.3 USD
76915-624 76915-626 76915-628
3-Fluoro-2-methoxy-5-methylbenzaldehyde ≥97%
3-Fluoro-2-methoxy-5-methylbenzaldehyde
Formula: C₉H₉FO₂
MW: 168.165 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24677397
CAS Number: 82129-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR