3-Methoxy-2-naphthoic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Methoxynaphthalene-2-carboxylic acid

36556-25G 36556-5G
76915-614EA 524.09 USD
76915-614 76915-616
3-Methoxy-2-naphthoic acid ≥97%
3-Methoxy-2-naphthoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₀O₃
MW: 202.2060 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00094168
CAS Number: 883-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR