4-(Difluoromethoxy)-3-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

36496-1G 36496-25G 36496-500MG 36496-5G
76915-598EA 409.51 USD
76915-598 76915-600 76915-602 76915-604
4-(Difluoromethoxy)-3-fluorobenzoic acid ≥97%
4-(Difluoromethoxy)-3-fluorobenzoic acid
Formula: C₈H₅F₃O₃
Storage Temperature: Refrigerator
MDL Number: MFCD14698507
CAS Number: 1214383-15-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR