3-Bromo-5-(methoxymethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

36136-1G 36136-25G 36136-500MG 36136-5G
76915-542EA 384.05 USD
76915-542 76915-544 76915-546 76915-548
3-Bromo-5-(methoxymethyl)benzoic acid ≥97%
3-Bromo-5-(methoxymethyl)benzoic acid
Formula: C₉H₉BrO₃
MW: 245.07 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24107173
CAS Number: 1536144-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR